(Chlorodifluoromethyl)trimethylsilane 97%

Lieferant: Apollo Scientific
Gefahr

PC10691-100MG PC10691-1G PC10691-250MG
APOSPC10691-100MGEA 263 EUR
APOSPC10691-100MG APOSPC10691-1G APOSPC10691-250MG
(Chlorodifluoromethyl)trimethylsilane 97%
(Chlorodifluoromethyl)trimethylsilane
Formel: C₄H₉ClF₂Si
Lagertemperatur: Kühlschrank
MDL: MFCD20486786
CAS-Nummer: 115262-00-5
UN: 1993
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR