Fluoromethyl 2-pyridyl sulphone 95%
Lieferant: Apollo Scientific
PC10695-1G
APOSPC10695-1GEA
186
EUR
APOSPC10695-1G
Fluoromethyl 2-pyridyl sulphone 95%
Fluoromethyl 2-pyridyl sulphone
Formel:
C₆H₆FNO₂S Molecular Weight: 175.181 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28100935 CAS-Nummer: 1365765-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...