(1S)-1-(3-Bromo-4-fluorophenyl)ethan-1-ol

Lieferant: Apollo Scientific
Achtung

PC10700-1G
APOSPC10700-1GEA 51.1 EUR
APOSPC10700-1G
(1S)-1-(3-Bromo-4-fluorophenyl)ethan-1-ol
(1S)-1-(3-Bromo-4-fluorophenyl)ethan-1-ol
Formel: C₈H₈BrFO
Molecular Weight: 219.051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09863797
CAS-Nummer: 929884-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR