4-Amino-3-chlor-5-nitrobenzotrifluorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1070D-100G
PC1070D-25G
PC1070D-5G
APOSPC1070D-100GEA
488
EUR
APOSPC1070D-100G
APOSPC1070D-25G
APOSPC1070D-5G
4-Amino-3-chlor-5-nitrobenzotrifluorid 98%
4-Amino-3-chlor-5-nitrobenzotrifluorid
Formel:
C₇H₄ClF₃N₂O₂ Schmelzpunkt: 66…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042153 CAS-Nummer: 57729-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...