5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile 97%

Lieferant: Apollo Scientific
Achtung

PC1070M-5G
APOSPC1070M-5GEA 108 EUR
APOSPC1070M-5G
5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile 97%
5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile
Formel: C₁₀H₇FN₄
Molecular Weight: 202.19 g/mol
Schmelzpunkt: 164…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00102729
CAS-Nummer: 51516-70-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR