2-Amino-3,5-dibromo-6-fluorobenzoic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3,5-Dibromo-6-fluoroanthranilic acid

PC1070N-1G
APOSPC1070N-1GEA 61.1 EUR
APOSPC1070N-1G
2-Amino-3,5-dibromo-6-fluorobenzoic acid 98%
2-Amino-3,5-dibromo-6-fluorobenzoic acid
Formel: C₇H₄Br₂FNO₂
Molecular Weight: 312,919 g/mol
Schmelzpunkt: 235…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042156
CAS-Nummer: 175135-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR