3-Amino-2-cyano-5-(4-fluorophenyl)thiophene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC1070Q-1G
APOSPC1070Q-1GEA
236
EUR
APOSPC1070Q-1G
3-Amino-2-cyano-5-(4-fluorophenyl)thiophene 95%
3-Amino-2-cyano-5-(4-fluorophenyl)thiophene
Formel:
C₁₁H₇FN₂S Schmelzpunkt: 199…200 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00210132 CAS-Nummer: 210356-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...