4-[(2-Aminoethyl)thio]-2-(trifluoromethyl)quinoline 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-{[2-(Trifluoromethyl)quinolin-4-yl]thio}ethylamine

PC1070V-1G PC1070V-10G
APOSPC1070V-1GEA 39.3 EUR
APOSPC1070V-1G APOSPC1070V-10G
4-[(2-Aminoethyl)thio]-2-(trifluoromethyl)quinoline 97%
4-[(2-Aminoethyl)thio]-2-(trifluoromethyl)quinoline
Formel: C₁₂H₁₁F₃N₂S
Molecular Weight: 272.289 g/mol
Schmelzpunkt: 97…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00174223
CAS-Nummer: 175203-50-6
UN: 3259
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR