4-(4-Fluorophenoxy)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
PC10711-1G
APOSPC10711-1GEA
319
EUR
APOSPC10711-1G
4-(4-Fluorophenoxy)benzyl bromide
4-(4-Fluorophenoxy)benzyl bromide
Formel:
C₁₃H₁₀BrFO Lagertemperatur: Raumtemperatur |
MDL:
MFCD15146807 CAS-Nummer: 83642-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...