5-Amino-2-(4-bromophenoxy)benzotrifluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Bromophenoxy)-3-(trifluoromethyl)aniline, 4-Amino-4'-bromo-2-(trifluoromethyl)diphenyl ether

PC10712-1G
APOSPC10712-1GEA 342 EUR
APOSPC10712-1G
5-Amino-2-(4-bromophenoxy)benzotrifluoride
5-Amino-2-(4-bromophenoxy)benzotrifluoride
Formel: C₁₃H₉BrF₃NO
Molecular Weight: 332,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987557
CAS-Nummer: 74338-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR