5-Fluoranthranilsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-fluorobenzoic acid
PC1071F-100G
APOSPC1071F-100GEA
97.3
EUR
APOSPC1071F-100G
5-Fluoranthranilsäure 97%
5-Fluoranthranilsäure
Formel:
C₇H₆FNO₂ Molecular Weight: 155.13 g/mol Siedepunkt: 305 Schmelzpunkt: 181…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055566 CAS-Nummer: 446-08-2 EINECS: 207-159-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...