3-Amino-2-(2-fluorphenoxy)pyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1076M-500MG
APOSPC1076M-500MGEA
72.5
EUR
APOSPC1076M-500MG
3-Amino-2-(2-fluorphenoxy)pyridin 98%
3-Amino-2-(2-fluorophenoxy)pyridine
Formel:
C₁₁H₉FN₂O Schmelzpunkt: 87…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067806 CAS-Nummer: 175135-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...