4-Amino-5-fluorchinazolin 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC1076T-1G
APOSPC1076T-1GEA
125
EUR
APOSPC1076T-1G
4-Amino-5-fluorchinazolin 95+%
4-Amino-5-fluoroquinazoline
Formel:
C₈H₆FN₃ Schmelzpunkt: 212…214 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153064 CAS-Nummer: 137553-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...