2-Bromo-6-methyl-4-(trifluoromethoxy)aniline

Lieferant: Apollo Scientific
Achtung

PC1084-1G
APOSPC1084-1GEA 357 EUR
APOSPC1084-1G
2-Bromo-6-methyl-4-(trifluoromethoxy)aniline
2-Bromo-6-methyl-4-(trifluoromethoxy)aniline
Formel: C₈H₇BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07368728
CAS-Nummer: 887266-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR