2-Fluor-5-(trifluormethoxy)benzylalkohol

Lieferant: Apollo Scientific

PC1093-5G
APOSPC1093-5GEA 453 EUR
APOSPC1093-5G
2-Fluor-5-(trifluormethoxy)benzylalkohol
2-Fluor-5-(trifluormethoxy)benzylalkohol
Formel: C₈H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115889
CAS-Nummer: 344411-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR