2-Fluor-5-(trifluormethoxy)benzylbromid

Lieferant: Apollo Scientific
Gefahr

PC1095-1G PC1095-5G
APOSPC1095-1GEA 0 EUR
APOSPC1095-1G APOSPC1095-5G
2-Fluor-5-(trifluormethoxy)benzylbromid
2-Fluor-5-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD04115890
CAS-Nummer: 86256-24-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR