4-Amino-2,3,5,6-tetrafluorbenzoesäure 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-2,3,5,6-tetrafluorobenzoic Acid

PC1098-1G PC1098-25G PC1098-5G
APOSPC1098-1GEA 29.3 EUR
APOSPC1098-1G APOSPC1098-25G APOSPC1098-5G
4-Amino-2,3,5,6-tetrafluorbenzoesäure 99%
4-Amino-2,3,5,6-tetrafluorbenzoesäure
Formel: C₇H₃F₄NO₂
Molecular Weight: 209.1 g/mol
Schmelzpunkt: 181…187 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00007647
CAS-Nummer: 944-43-4
EINECS: 213-409-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR