3-Amino-4-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC110014-1G PC110014-250MG
APOSPC110014-1GEA 0 EUR
APOSPC110014-1G APOSPC110014-250MG
3-Amino-4-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
3-Amino-4-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
Formel: C₁₀H₆F₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD29058792
CAS-Nummer: 1648807-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR