5-Amino-2-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC110019-1G PC110019-250MG
APOSPC110019-1GEA 0 EUR
APOSPC110019-1G APOSPC110019-250MG
5-Amino-2-(trifluoromethyl)benzoic acid
5-Amino-2-(trifluoromethyl)benzoic acid
Formel: C₈H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28505050
CAS-Nummer: 1245457-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR