3-Amino-2-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC110022-1G PC110022-250MG
APOSPC110022-1GEA 0 EUR
APOSPC110022-1G APOSPC110022-250MG
3-Amino-2-(trifluoromethyl)benzoic acid
3-Amino-2-(trifluoromethyl)benzoic acid
Formel: C₈H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22480979
CAS-Nummer: 1369896-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR