3-(4-Fluoro-1-naphthyl)-1H-pyrazole
Lieferant: Apollo Scientific
PC110028-1G
APOSPC110028-1GEA
183
EUR
APOSPC110028-1G
3-(4-Fluoro-1-naphthyl)-1H-pyrazole
3-(4-Fluoro-1-naphthyl)-1H-pyrazole
Formel:
C₁₃H₉FN₂ Molecular Weight: 212,222 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407406 CAS-Nummer: 1019011-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...