3-[4-(2,4-Difluorophenoxy)phenyl]-1H-pyrazole

Lieferant: Apollo Scientific

PC110032-1G
APOSPC110032-1GEA 183 EUR
APOSPC110032-1G
3-[4-(2,4-Difluorophenoxy)phenyl]-1H-pyrazole
3-[4-(2,4-Difluorophenoxy)phenyl]-1H-pyrazole
Formel: C₁₅H₁₀F₂N₂O
Molecular Weight: 272,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06409160
CAS-Nummer: 861213-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR