8-Bromo-6-fluoro-4-hydroxyquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

PC110035-1G
APOSPC110035-1GEA 65.4 EUR
APOSPC110035-1G
8-Bromo-6-fluoro-4-hydroxyquinoline-3-carboxylic acid
8-Bromo-6-fluoro-4-hydroxyquinoline-3-carboxylic acid
Formel: C₁₀H₅BrFNO₃
Molecular Weight: 286,054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09861322
CAS-Nummer: 1019016-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR