Ethyl 8-bromo-5-fluoro-4-hydroxyquinoline-3-carboxylate
Lieferant: Apollo Scientific
PC110038-1G
PC110038-250MG
APOSPC110038-1GEA
284
EUR
APOSPC110038-1G
APOSPC110038-250MG
Ethyl 8-bromo-5-fluoro-4-hydroxyquinoline-3-carboxylate
Ethyl 8-bromo-5-fluoro-4-hydroxyquinoline-3-carboxylate
Formel:
C₁₂H₉BrFNO₃ Molecular Weight: 314.107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16036881 CAS-Nummer: 655236-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...