4-Amino-3-(trifluormethoxy)benzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1100BD-1G
PC1100BD-25G
PC1100BD-5G
APOSPC1100BD-1GEA
0
EUR
APOSPC1100BD-1G
APOSPC1100BD-25G
APOSPC1100BD-5G
4-Amino-3-(trifluormethoxy)benzoesäure 97%
4-Amino-3-(trifluoromethoxy)benzoic acid
Formel:
C₈H₆F₃NO₃ Schmelzpunkt: 168…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205143 CAS-Nummer: 175278-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...