4-Amino-3-(trifluormethoxy)benzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1100BF-10G
APOSPC1100BF-10GEA
178
EUR
APOSPC1100BF-10G
4-Amino-3-(trifluormethoxy)benzonitril 97%
4-Amino-3-(trifluormethoxy)benzonitril
Formel:
C₈H₅F₃N₂O Schmelzpunkt: 71…73 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00204177 CAS-Nummer: 175278-23-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...