4-Amino-2-(trifluoromethyl)acetanilide 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1100CA-1G
APOSPC1100CA-1GEA
116
EUR
APOSPC1100CA-1G
4-Amino-2-(trifluoromethyl)acetanilide 98%
4-Amino-2-(trifluoromethyl)acetanilide
Formel:
C₉H₉F₃N₂O Schmelzpunkt: 137…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052847 CAS-Nummer: 134514-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...