2-(1-Pyrrolidinyl)-5-(trifluoromethyl)aniline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[2-Amino-4-(trifluoromethyl)phenyl]pyrrolidine

PC1100F-500MG
APOSPC1100F-500MGEA 21.9 EUR
APOSPC1100F-500MG
2-(1-Pyrrolidinyl)-5-(trifluoromethyl)aniline 97%
2-(1-Pyrrolidinyl)-5-(trifluoromethyl)aniline
Formel: C₁₁H₁₃F₃N₂
Molecular Weight: 230,23 g/mol
Schmelzpunkt: 33…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042162
CAS-Nummer: 133184-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR