Ethyl 8-fluoro-4-hydroxy-6-iodoquinoline-3-carboxylate ≥98%

Lieferant: Apollo Scientific

PC110107-1G
APOSPC110107-1GEA 71.2 EUR
APOSPC110107-1G
Ethyl 8-fluoro-4-hydroxy-6-iodoquinoline-3-carboxylate ≥98%
Ethyl 8-fluoro-4-hydroxy-6-iodoquinoline-3-carboxylate
Formel: C₁₂H₉FINO₃
Molecular Weight: 361,108 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12922926
CAS-Nummer: 228728-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR