4-(Trifluormethyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(Trifluormethyl)phenyl)metaborsäure, Dihydroxy(4-(trifluormethyl)phenyl)boran, 4-(Trifluormethyl)benzolboronsäure

PC1111-100G PC1111-25G PC1111-5G
APOSPC1111-100GEA 109 EUR
APOSPC1111-100G APOSPC1111-25G APOSPC1111-5G
4-(Trifluormethyl)phenylboronsäure 98%
4-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
Molecular Weight: 189.93 g/mol
Schmelzpunkt: 235…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151855
CAS-Nummer: 128796-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR