4-Chlor-3-(trifluormethyl)phenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC11111-25G
PC11111-5G
PC11111-1G
APOSPC11111-25GEA
267
EUR
APOSPC11111-25G
APOSPC11111-5G
APOSPC11111-1G
4-Chlor-3-(trifluormethyl)phenylboronsäure 95%
4-Chlor-3-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BClF₃O₂ Schmelzpunkt: 233…236 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094999 CAS-Nummer: 176976-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...