2-Methoxy-5-(trifluormethyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methoxy-5-(trifluoromethyl)benzeneboronic acid
PC11116-5G
APOSPC11116-5GEA
105
EUR
APOSPC11116-5G
2-Methoxy-5-(trifluormethyl)phenylboronsäure
2-Methoxy-5-(trifluormethyl)phenylboronsäure
Formel:
C₈H₈BF₃O₃ Molecular Weight: 219.95 g/mol Schmelzpunkt: 144…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039222 CAS-Nummer: 240139-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...