1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione

Lieferant: Apollo Scientific

PC11122-1G
APOSPC11122-1GEA 161 EUR
APOSPC11122-1G
1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione
1-[3-(Trifluoromethyl)phenyl]-1H-pyrrole-2,5-dione
Formel: C₁₁H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00439448
CAS-Nummer: 53629-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR