3-Amino-3'-fluorobiphenyl hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3-Fluorophenyl)aniline hydrochloride, 3'-Fluoro-[1,1'-biphenyl]-3-amine hydrochloride

PC11126-1G
APOSPC11126-1GEA 105 EUR
APOSPC11126-1G
3-Amino-3'-fluorobiphenyl hydrochloride
3-Amino-3'-fluorobiphenyl hydrochloride
Formel: C₁₂H₁₀FN·ClH
Molecular Weight: 223,674 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08443612
CAS-Nummer: 1210215-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR