2'-Fluoro-[1,1'-biphenyl]-4-carboxaldehyde 95+%
Lieferant: Apollo Scientific
Synonyme:
2'-Fluoro-[1,1'-biphenyl]-4-carbaldehyde
PC11131-1G
APOSPC11131-1GEA
404
EUR
APOSPC11131-1G
2'-Fluoro-[1,1'-biphenyl]-4-carboxaldehyde 95+%
2'-Fluoro-[1,1'-biphenyl]-4-carboxaldehyde
Formel:
C₁₃H₉FO Molecular Weight: 200,208 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424638 CAS-Nummer: 57592-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...