4'-Fluoro-biphenyl-3-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
PC11136-1G
APOSPC11136-1GEA
121
EUR
APOSPC11136-1G
4'-Fluoro-biphenyl-3-carbaldehyde
4'-Fluoro-biphenyl-3-carbaldehyde
Formel:
C₁₃H₉FO Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631878 CAS-Nummer: 164334-74-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...