3'-Fluorbiphenyl-3-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC11140-250MG
APOSPC11140-250MGEA
62.5
EUR
APOSPC11140-250MG
3'-Fluorbiphenyl-3-carbonsäure
3'-Fluorbiphenyl-3-carbonsäure
Formel:
C₁₃H₉FO₂ Molecular Weight: 216,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424608 CAS-Nummer: 168619-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...