4'-Fluorbiphenyl-3-carbonsäure 95%

Lieferant: Apollo Scientific
Achtung

PC11143-5G
APOSPC11143-5GEA 142 EUR
APOSPC11143-5G
4'-Fluorbiphenyl-3-carbonsäure 95%
4'-Fluorbiphenyl-3-carbonsäure
Formel: C₁₃H₉FO₂
Molecular Weight: 216.21
Lagertemperatur: Raumtemperatur
MDL: MFCD00452724
CAS-Nummer: 10540-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR