3'-Fluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(3-Fluorophenyl)salicyclic acid, 5-(3-Fluorophenyl)-2-hydroxybenzoic acid

PC11149-1G
APOSPC11149-1GEA 667 EUR
APOSPC11149-1G
3'-Fluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
3'-Fluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Formel: C₁₃H₉FO₃
Molecular Weight: 232,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039228
CAS-Nummer: 22494-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR