5-(3-Fluorophenyl)-2-furoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(3-Fluorophenyl)furan-2-carboxylic acid
PC11152-100MG
APOSPC11152-100MGEA
34.8
EUR
APOSPC11152-100MG
5-(3-Fluorophenyl)-2-furoic acid
5-(3-Fluorophenyl)-2-furoic acid
Formel:
C₁₁H₇FO₃ Molecular Weight: 206,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039229 CAS-Nummer: 54022-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...