4-Hydroxy-2'-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-5-[2-(trifluoromethyl)phenyl]benzoic acid, 5-[2-(Trifluoromethyl)phenyl]salicylic acid

PC11157-500MG
APOSPC11157-500MGEA 105 EUR
APOSPC11157-500MG
4-Hydroxy-2'-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid
4-Hydroxy-2'-(trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid
Formel: C₁₄H₉F₃O₃
Molecular Weight: 282,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039233
CAS-Nummer: 400744-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR