4,5-Difluor-2-hydroxybenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
PC1116-250MG
APOSPC1116-250MGEA
161
EUR
APOSPC1116-250MG
4,5-Difluor-2-hydroxybenzonitril
4,5-Difluor-2-hydroxybenzonitril
Formel:
C₇H₃F₂NO Molecular Weight: 155,10 g/mol Schmelzpunkt: 161…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08275287 CAS-Nummer: 186590-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...