3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-(Trifluoromethyl)-[1,1'-biphenyl]-2-amine hydrochloride

PC11163-100MG
APOSPC11163-100MGEA 69.8 EUR
APOSPC11163-100MG
3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine 96%
3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine
Formel: C₁₃H₁₀F₃N
Molecular Weight: 237,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424654
CAS-Nummer: 365-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR