3-Amino-3'-(trifluoromethyl)biphenyl hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[3-(Trifluoromethyl)phenyl]aniline hydrochloride, 3'-(Trifluoromethyl)-[1,1'-biphenyl]-3-amine hydrochloride

PC11165-1G
APOSPC11165-1GEA 241 EUR
APOSPC11165-1G
3-Amino-3'-(trifluoromethyl)biphenyl hydrochloride
3-Amino-3'-(trifluoromethyl)biphenyl hydrochloride
Formel: C₁₃H₁₀F₃N·ClH
Molecular Weight: 273,681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739426
CAS-Nummer: 811842-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR