4-Amino-4'-(trifluoromethyl)biphenyl hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[4-(Trifluoromethyl)phenyl]aniline hydrochloride, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-amine hydrochloride, 4-(4-Aminophenyl)benzotrifluoride hydrochloride

PC11169-100MG
APOSPC11169-100MGEA 59.6 EUR
APOSPC11169-100MG
4-Amino-4'-(trifluoromethyl)biphenyl hydrochloride
4-Amino-4'-(trifluoromethyl)biphenyl hydrochloride
Formel: C₁₃H₁₀F₃N·ClH
Molecular Weight: 273,681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445324
CAS-Nummer: 1209254-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR