4'-(Trifluormethyl)-4-biphenylcarbaldehyd

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxaldehyde, 4-(4-Trifluoromethylphenyl)benzaldehyde, 4'-Trifluoromethyl-biphenyl-4-carbaldehyde, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

PC11172-1G
APOSPC11172-1GEA 27.7 EUR
APOSPC11172-1G
4'-(Trifluormethyl)-4-biphenylcarbaldehyd
4'-Trifluoromethylbiphenyl-4-carbaldehyde
Formel: C₁₄H₉F₃O
Molecular Weight: 250,2200 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01862519
CAS-Nummer: 90035-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR