3'-(Trifluoromethyl)biphenyl-4-carboxaldehyde

Lieferant: Apollo Scientific
Gefahr

PC11176-1G PC11176-5G
APOSPC11176-1GEA 132 EUR
APOSPC11176-1G APOSPC11176-5G
3'-(Trifluoromethyl)biphenyl-4-carboxaldehyde
3'-(Trifluoromethyl)biphenyl-4-carboxaldehyde
Formel: C₁₄H₉F₃O
Molecular Weight: 250.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424645
CAS-Nummer: 100036-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR