3'-(Trifluormethyl)biphenyl-4-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC11178-1G
PC11178-250MG
PC11178-5G
APOSPC11178-1GEA
118
EUR
APOSPC11178-1G
APOSPC11178-250MG
APOSPC11178-5G
3'-(Trifluormethyl)biphenyl-4-carbonsäure
3'-(Trifluormethyl)biphenyl-4-carbonsäure
Formel:
C₁₄H₉F₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424607 CAS-Nummer: 195457-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...