3'-(Trifluormethyl)biphenyl-4-carbonsäure

Lieferant: Apollo Scientific
Achtung

PC11178-1G PC11178-250MG PC11178-5G
APOSPC11178-1GEA 118 EUR
APOSPC11178-1G APOSPC11178-250MG APOSPC11178-5G
3'-(Trifluormethyl)biphenyl-4-carbonsäure
3'-(Trifluormethyl)biphenyl-4-carbonsäure
Formel: C₁₄H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03424607
CAS-Nummer: 195457-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR