4'-(Trifluormethyl)biphenyl-4-carbonsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC11180-1G
PC11180-5G
APOSPC11180-1GEA
75.4
EUR
APOSPC11180-1G
APOSPC11180-5G
4'-(Trifluormethyl)biphenyl-4-carbonsäure 98%
4'-(Trifluormethyl)biphenyl-4-carbonsäure
Formel:
C₁₄H₉F₃O₂ Schmelzpunkt: 321…324 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424606 CAS-Nummer: 195457-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...